siRNA Details
Gene Symbol: Hcrtr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013074.1 | NP_037206.1 | 4 | 785 | ||
XM_008766215.1 | XP_008764437.1 | 4 | 1145 | |||
GenBank mRNA | AF041246.1 | - | 4 | 785 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
