siRNA Details
Gene Symbol: G6pc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013098.2 | NP_037230.2 | 5 | 1019 | ||
GenBank mRNA | BC090067.1 | - | 5 | 1019 | 7109006 | |
D78592.1 | - | 5 | 1031 | |||
L37333.1 | - | 5 | 983 | |||
U07993.1 | - | 5 | 981 |
