siRNA Details
Gene Symbol: Igfbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013122.2 | NP_037254.2 | 3, 4 | 825 | ||
GenBank mRNA | BC092570.1 | - | 3, 4 | 825 | 7308779 | |
FQ212531.1 | - | 3, 4 | 844 | |||
J04486.1 | - | 3, 4 | 1017 | |||
M31672.1 | - | 3, 4 | 853 |
