siRNA Details
Gene Symbol: Scn5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160162.1 | NP_001153634.1 | 22, 23 | 4287 | ||
NM_013125.2 | NP_037257.1 | 23, 24 | 4446 | |||
XM_006244076.2 | XP_006244138.1 | 23, 24 | 4616 | |||
XM_008766670.1 | XP_008764892.1 | 23, 24 | 5644 | |||
XM_008766671.1 | XP_008764893.1 | 22, 23 | 5485 | |||
GenBank mRNA | AF353637.1 | - | 21, 22 | 4090 | ||
M27902.1 | - | 23, 24 | 4446 |
