siRNA Details
Gene Symbol: Anxa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013132.1 | NP_037264.1 | 5 | 304 | ||
XM_006232268.2 | XP_006232330.1 | 5 | 439 | |||
GenBank mRNA | AF051895.1 | - | 5 | 359 | ||
BC081855.1 | - | 5 | 357 | 7192977 | ||
FQ213209.1 | - | 6 | 434 | |||
FQ214176.1 | - | 5 | 334 | |||
FQ229127.1 | - | 5 | 292 | |||
M21730.1 | - | 5 | 304 |
