siRNA Details
Gene Symbol: Ccne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100821.1 | NP_001094291.1 | 6 | 522 | ||
XM_006228904.2 | XP_006228966.1 | 6 | 674 | |||
GenBank mRNA | BC162008.1 | - | 6 | 549 | 9090267 | |
D14015.1 | - | 6 | 539 | |||
EV771022.1 | - | 6 | 556 |
