siRNA Details
Gene Symbol: Syt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019122.1 | NP_061995.1 | 8, 9 | 1814 | ||
XM_006228978.2 | XP_006229040.1 | 9, 10 | 2272 | |||
XM_006228979.2 | XP_006229041.1 | 9, 10 | 2277 | |||
XM_008759324.1 | XP_008757546.1 | 10, 11 | 2002 | |||
XM_008759325.1 | XP_008757547.1 | 9, 10 | 1906 | |||
GenBank mRNA | AF375464.1 | - | 7, 8 | 1712 | ||
D28512.1 | - | 8, 9 | 1814 | |||
EU441216.1 | - | 7, 8 | 1717 |
