siRNA Details
Gene Symbol: Frmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271054.1 | NP_001257983.1 | 8, 9 | 1012 | ||
XM_006240151.2 | XP_006240213.1 | 8, 9 | 1062 | |||
XM_006240152.2 | XP_006240214.1 | 9, 10 | 1142 | |||
XM_006240153.2 | XP_006240215.1 | 8, 9 | 1486 | |||
GenBank mRNA | AF441249.1 | - | 7, 8 | 789 |
