siRNA Details
Gene Symbol: Frmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271054.1 | NP_001257983.1 | 14 | 2046 | ||
XM_006240151.2 | XP_006240213.1 | 14 | 2096 | |||
XM_006240152.2 | XP_006240214.1 | 15 | 2176 | |||
XM_006240153.2 | XP_006240215.1 | 14 | 2520 |
