siRNA Details
Gene Symbol: Gjc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085381.1 | NP_001078850.1 | 1219 | |||
GenBank mRNA | AF536559.4 | - | 812 | |||
BC166405.1 | - | 1104 | 9021864 |

More Information
Other Species Targeted

Gene Ontology Categories:
