siRNA Details
Gene Symbol: Rnf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004075.1 | NP_001004075.1 | 3, 4 | 738 | ||
XM_006249360.2 | XP_006249422.1 | 3, 4 | 1271 | |||
XM_006249361.2 | XP_006249423.1 | 4, 5 | 948 | |||
XM_008769214.1 | XP_008767436.1 | 3, 4 | 656 | |||
GenBank mRNA | AY157968.1 | - | 2, 3 | 643 | ||
BC079044.1 | - | 3, 4 | 738 | 7115027 |
