siRNA Details
Gene Symbol: Hsbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173119.2 | NP_775142.1 | 2 | 162 | ||
GenBank mRNA | AF522937.1 | - | 2 | 109 | ||
BC058131.1 | - | 2 | 147 | 6919748 | ||
EX491168.1 | - | 2 | 162 | |||
FQ211891.1 | - | 2 | 130 | |||
FQ212621.1 | - | 2 | 134 | |||
FQ213728.1 | - | 2 | 104 | |||
FQ216175.1 | - | 2 | 141 | |||
FQ229155.1 | - | 2 | 147 | |||
FQ229754.1 | - | 2 | 121 | |||
FQ230047.1 | - | 2 | 119 | |||
FQ230722.1 | - | 2 | 121 | |||
FQ233773.1 | - | 2 | 120 |
