siRNA Details
Gene Symbol: Cdk5rap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173134.2 | NP_775157.2 | 23, 24 | 3307 | ||
XM_006238273.2 | XP_006238335.1 | 23, 24 | 3543 | |||
XM_006238274.2 | XP_006238336.1 | 23, 24 | 3537 | |||
XM_006238276.2 | XP_006238338.1 | 22, 23 | 3445 | |||
XM_006238277.2 | XP_006238339.1 | 22, 23 | 3439 | |||
XM_006238278.2 | XP_006238340.1 | 23, 24 | 3541 | |||
XM_006238279.2 | XP_006238341.1 | 23, 24 | 3534 | |||
XM_006238280.2 | XP_006238342.1 | 23, 24 | 3541 | |||
XM_008763760.1 | XP_008761982.1 | 23, 24 | 3543 | |||
GenBank mRNA | CB806103.1 | - | 2, 3 | 294 | ||
JX524852.1 | - | 23, 24 | 3151 |
