siRNA Details
Gene Symbol: Cdk5rap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173134.2 | NP_775157.2 | 24 | 3337 | ||
XM_006238273.2 | XP_006238335.1 | 24 | 3573 | |||
XM_006238274.2 | XP_006238336.1 | 24 | 3567 | |||
XM_006238276.2 | XP_006238338.1 | 23 | 3475 | |||
XM_006238277.2 | XP_006238339.1 | 23 | 3469 | |||
XM_006238278.2 | XP_006238340.1 | 24 | 3571 | |||
XM_006238279.2 | XP_006238341.1 | 24 | 3564 | |||
XM_006238280.2 | XP_006238342.1 | 24 | 3571 | |||
XM_008763760.1 | XP_008761982.1 | 24 | 3573 | |||
XM_008763761.1 | XP_008761983.1 | 23 | 3442 | |||
GenBank mRNA | JX524852.1 | - | 24 | 3181 |
