siRNA Details
Gene Symbol: Dlgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276304.1 | NP_001263233.1 | 4, 5 | 1507 | ||
XM_008763989.1 | XP_008762211.1 | 5, 6 | 2809 | |||
GenBank mRNA | AY530298.1 | - | 3, 4 | 1370 | ||
AY530299.1 | - | 3, 4 | 1370 | |||
U67139.1 | - | 4, 5 | 1474 |
