siRNA Details
Gene Symbol: Vps54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173147.1 | NP_775170.1 | 19 | 2641 | ||
GenBank mRNA | AJ010392.3 | - | 19 | 2641 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vps54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173147.1 | NP_775170.1 | 19 | 2641 | ||
GenBank mRNA | AJ010392.3 | - | 19 | 2641 |