siRNA Details
Gene Symbol: Havcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173149.2 | NP_775172.1 | 2 | 860 | ||
XM_006246163.2 | XP_006246225.1 | 3 | 505 | |||
XM_008767665.1 | XP_008765887.1 | 4 | 557 | |||
XM_008767666.1 | XP_008765888.1 | 3 | 1142 | |||
GenBank mRNA | AF035963.1 | - | 2 | 861 | ||
BC061820.1 | - | 3 | 300 | 5599247 |
