siRNA Details
Gene Symbol: Elavl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238385.2 | XP_006238447.1 | 7 | 986 | ||
XM_006238388.2 | XP_006238450.1 | 6 | 789 | |||
XM_006238389.2 | XP_006238451.1 | 6 | 1326 | |||
XM_006238391.2 | 6 | 1326 | ||||
XM_006238395.2 | XP_006238457.1 | 5 | 717 | |||
XM_008763848.1 | XP_008762070.1 | 5 | 1032 | |||
XM_008763849.1 | XP_008762071.1 | 5 | 1032 | |||
XM_008763850.1 | XP_008762072.1 | 5 | 695 | |||
GenBank mRNA | AB098713.1 | - | 6 | 1250 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001177883, NM_207686, NM_010486, NM_207685, U29088, and others.
Gene Ontology Categories:
