siRNA Details
Gene Symbol: Clec16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767505.1 | XP_008765727.1 | 24 | 3057 | ||
XM_008767506.1 | XP_008765728.1 | 23 | 3051 | |||
XM_008767507.1 | XP_008765729.1 | 24 | 3009 | |||
XM_008767508.1 | XP_008765730.1 | 23 | 3003 | |||
XM_008767509.1 | XP_008765731.1 | 23 | 3003 |
