siRNA Details
Gene Symbol: Rogdi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024864.1 | NP_001020035.1 | 10 | 991 | ||
GenBank mRNA | BC083789.1 | - | 8 | 1422 | ||
BC098001.1 | - | 10 | 991 | 7453517 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rogdi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024864.1 | NP_001020035.1 | 10 | 991 | ||
GenBank mRNA | BC083789.1 | - | 8 | 1422 | ||
BC098001.1 | - | 10 | 991 | 7453517 |