siRNA Details
Gene Symbol: Cldn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011889.1 | NP_001011889.1 | 1 | 986 | ||
GenBank mRNA | BC087664.1 | - | 1 | 986 | 7315462 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cldn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011889.1 | NP_001011889.1 | 1 | 986 | ||
GenBank mRNA | BC087664.1 | - | 1 | 986 | 7315462 |