siRNA Details
Gene Symbol: Kremen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105767.1 | NP_001099237.1 | 6 | 709 | ||
XM_008767550.1 | XP_008765772.1 | 5 | 932 | |||
XM_008767551.1 | XP_008765773.1 | 5 | 932 | |||
XM_008767552.1 | XP_008765774.1 | 5 | 932 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028416, BC125438, AK037113, AJ457192, BC120532.
Gene Ontology Categories:
