siRNA Details
Gene Symbol: Mrps34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105771.1 | NP_001099241.1 | 3 | 486 | ||
GenBank mRNA | FQ228234.1 | - | 3 | 445 | ||
FQ230451.1 | - | 2 | 569 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mrps34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105771.1 | NP_001099241.1 | 3 | 486 | ||
GenBank mRNA | FQ228234.1 | - | 3 | 445 | ||
FQ230451.1 | - | 2 | 569 |