siRNA Details
Gene Symbol: Gnptg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100493.1 | NP_001093963.1 | 4 | 211 | ||
XM_006245935.2 | XP_006245997.1 | 4 | 228 | |||
GenBank mRNA | BC098716.1 | - | 4 | 193 | ||
BC158818.1 | - | 4 | 211 | 7374209 | ||
FQ213320.1 | - | 4 | 218 | |||
FQ213586.1 | - | 4 | 176 | |||
FQ213702.1 | - | 4 | 210 |
