siRNA Details
Gene Symbol: Rpusd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105774.1 | NP_001099244.1 | 6 | 700 | ||
XM_006245937.2 | XP_006245999.1 | 6 | 755 | |||
GenBank mRNA | BC161966.1 | - | 6 | 698 | 9028415 | |
FQ213277.1 | - | 6 | 621 |
