siRNA Details
Gene Symbol: B9d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105786.1 | NP_001099256.1 | 7 | 559 | ||
GenBank mRNA | BC169130.1 | - | 8 | 649 | 6889289 |
Gene Symbol: LOC100911746 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750843.3 | XP_003750891.1 | 7 | 635 |

More Information
Other Species Targeted

Gene Ontology Categories:
