siRNA Details
Gene Symbol: Ctdnep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100494.1 | NP_001093964.1 | 3 | 396 | ||
XM_006246643.2 | XP_006246705.1 | 4 | 521 | |||
XM_006246644.2 | XP_006246706.1 | 4 | 521 | |||
XM_008767804.1 | XP_008766026.1 | 3 | 425 | |||
GenBank mRNA | BC087638.1 | - | 1 | 30 | ||
BC107474.1 | - | 2 | 57 | |||
BU671232.1 | - | 3 | 396 |
