siRNA Details
Gene Symbol: Wfikkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247173.2 | XP_006247235.1 | 2 | 1303 | ||
XM_220855.8 | XP_220855.1 | 2 | 2152 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wfikkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247173.2 | XP_006247235.1 | 2 | 1303 | ||
XM_220855.8 | XP_220855.1 | 2 | 2152 |