siRNA Details
Gene Symbol: Nkiras2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105839.1 | NP_001099309.1 | 2 | 215 | ||
XM_006247274.2 | XP_006247336.1 | 2 | 166 | |||
XM_008767974.1 | XP_008766196.1 | 2 | 172 | |||
GenBank mRNA | BC168163.1 | - | 2 | 109 | 8366401 |
