siRNA Details
Gene Symbol: Nkiras2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105839.1 | NP_001099309.1 | 3 | 370 | ||
XM_006247274.2 | XP_006247336.1 | 3 | 321 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nkiras2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105839.1 | NP_001099309.1 | 3 | 370 | ||
XM_006247274.2 | XP_006247336.1 | 3 | 321 |