siRNA Details
Gene Symbol: Eif4g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248581.2 | XP_006248643.1 | 14 | 2262 | ||
XM_006248582.1 | XP_006248644.1 | 14 | 2251 | |||
XM_006248583.1 | XP_006248645.1 | 14 | 2254 | |||
XM_006248584.2 | XP_006248646.1 | 15 | 2329 | |||
XM_006248585.2 | XP_006248647.1 | 14 | 2270 | |||
XM_008768835.1 | XP_008767057.1 | 15 | 2326 |
Gene Symbol: LOC100911693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760888.1 | XP_008759110.1 | 1 | 1522 |
