siRNA Details
Gene Symbol: Eif4g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248581.2 | XP_006248643.1 | 22, 23 | 3463 | ||
XM_006248582.1 | XP_006248644.1 | 22, 23 | 3452 | |||
XM_006248583.1 | XP_006248645.1 | 22, 23 | 3455 | |||
XM_006248584.2 | XP_006248646.1 | 23, 24 | 3530 | |||
XM_006248585.2 | XP_006248647.1 | 22, 23 | 3471 | |||
XM_008768835.1 | XP_008767057.1 | 23, 24 | 3527 |
Gene Symbol: LOC100912571 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248555.1 | XP_006248617.1 | 254 |
