siRNA Details
Gene Symbol: Lpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013864.1 | NP_001013886.1 | 10 | 1695 | ||
XM_006248505.2 | XP_006248567.1 | 13 | 2132 | |||
XM_006248507.2 | XP_006248569.1 | 7 | 9598 | |||
GenBank mRNA | BC083627.1 | - | 10 | 1695 | 7134282 | |
BC083890.1 | - | 9 | 1633 | 7132481 |
