siRNA Details
Gene Symbol: Ropn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025628.1 | NP_001020799.1 | 4 | 475 | ||
XM_006248431.2 | XP_006248493.1 | 4 | 418 | |||
GenBank mRNA | BC099132.1 | - | 4 | 475 | 7453834 |
