siRNA Details
Gene Symbol: Mylk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105874.2 | NP_001099344.2 | 31 | 5652 | ||
XM_006248433.2 | XP_006248495.1 | 31 | 5960 | |||
XM_006248434.2 | XP_006248496.1 | 31 | 5742 | |||
XM_008768711.1 | XP_008766933.1 | 31 | 5958 | |||
XM_008768712.1 | XP_008766934.1 | 15 | 2372 |
