siRNA Details
Gene Symbol: Nit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034126.1 | NP_001029298.1 | 7, 8 | 634 | ||
GenBank mRNA | BC082034.1 | - | 6, 7 | 1656 | ||
BC100637.1 | - | 7, 8 | 634 | 5621093 | ||
FQ209436.1 | - | 7, 8 | 642 | |||
FQ219054.1 | - | 4, 5 | 566 | |||
FQ219183.1 | - | 7, 8 | 640 | |||
FQ229934.1 | - | 7, 8 | 646 |
