siRNA Details
Gene Symbol: Pigp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099758.1 | NP_001093228.1 | 5 | 471 | ||
XM_006248042.2 | XP_006248104.1 | 4 | 542 | |||
XM_006248043.2 | XP_006248105.1 | 5 | 561 | |||
GenBank mRNA | AA926031.1 | - | 1 | 81 | ||
BC166404.1 | - | 5 | 561 | 9099247 | ||
CF978233.1 | - | 5 | 436 |
