siRNA Details
Gene Symbol: Mis18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127523.2 | NP_001120995.2 | 2, 3 | 392 | ||
GenBank mRNA | BC167102.1 | - | 2, 3 | 392 | 8373935 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mis18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127523.2 | NP_001120995.2 | 2, 3 | 392 | ||
GenBank mRNA | BC167102.1 | - | 2, 3 | 392 | 8373935 |