siRNA Details
Gene Symbol: Dhrsx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105914.1 | NP_001099384.1 | 4 | 325 | ||
XM_006248993.2 | XP_006249055.1 | 4 | 376 | |||
XM_006248995.2 | XP_006249057.1 | 4 | 377 | |||
XM_006248997.1 | XP_006249059.1 | 4 | 620 | |||
GenBank mRNA | BC167053.1 | - | 4 | 304 | 7110020 |
