siRNA Details
Gene Symbol: Fis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105919.1 | NP_001099389.1 | 2 | 339 | ||
XM_006249121.2 | XP_006249183.1 | 2 | 405 | |||
XM_006249122.2 | XP_006249184.1 | 2 | 359 | |||
XM_006249123.2 | XP_006249185.1 | 2 | 385 | |||
GenBank mRNA | BC167057.1 | - | 2 | 352 | 9098277 | |
FQ213833.1 | - | 2 | 165 | |||
FQ222246.1 | - | 2 | 151 | |||
FQ224250.1 | - | 2 | 162 | |||
FQ232488.1 | - | 2 | 151 |
