siRNA Details
Gene Symbol: Tmem248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004204.2 | NP_001004204.1 | 7 | 1017 | ||
XM_006249230.2 | XP_006249292.1 | 4 | 719 | |||
XM_006249231.2 | XP_006249293.1 | 4 | 640 | |||
GenBank mRNA | BC079161.1 | - | 4 | 691 | 7129411 | |
BC090318.1 | - | 7 | 1017 | 7113430 |
