siRNA Details
Gene Symbol: Tmem248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004204.2 | NP_001004204.1 | 6 | 986 | ||
XM_006249230.2 | XP_006249292.1 | 3 | 688 | |||
XM_006249231.2 | XP_006249293.1 | 3 | 609 | |||
GenBank mRNA | BC079161.1 | - | 3 | 660 | 7129411 | |
BC090318.1 | - | 6 | 986 | 7113430 |
