siRNA Details
Gene Symbol: Tesc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249426.2 | XP_006249488.1 | 9 | 820 | ||
XM_213790.9 | XP_213790.6 | 8 | 721 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tesc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249426.2 | XP_006249488.1 | 9 | 820 | ||
XM_213790.9 | XP_213790.6 | 8 | 721 |