siRNA Details
Gene Symbol: Wdr83os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105947.1 | NP_001099417.1 | 3 | 327 | ||
XM_006255237.1 | XP_006255299.1 | 4 | 556 | |||
GenBank mRNA | BC166543.1 | - | 4 | 432 | 7300989 | |
FQ225008.1 | - | 4 | 561 |
