siRNA Details
Gene Symbol: Cdk18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100506.1 | NP_001093976.1 | 4 | 357 | ||
XM_006249750.2 | XP_006249812.1 | 5 | 444 | |||
XM_006249751.2 | XP_006249813.1 | 5 | 505 | |||
XM_006249752.2 | XP_006249814.1 | 5 | 965 | |||
XM_006249753.2 | XP_006249815.1 | 5 | 478 | |||
XM_008769441.1 | XP_008767663.1 | 5 | 783 | |||
GenBank mRNA | AB005541.1 | - | 5 | 437 | ||
BC082045.1 | - | 4 | 357 | 7131722 |
