siRNA Details
Gene Symbol: Chit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079689.2 | NP_001073157.1 | 4 | 776 | ||
NM_001270846.1 | NP_001257775.1 | 5 | 425 | |||
NM_001270847.1 | NP_001257776.1 | 4 | 324 | |||
NM_001270848.1 | NP_001257777.1 | 4 | 409 | |||
XM_008769542.1 | XP_008767764.1 | 3 | 705 | |||
GenBank mRNA | DQ286232.1 | - | 3 | 302 | ||
DQ923316.1 | - | 3 | 247 | |||
DQ923317.1 | - | 4 | 409 | |||
DQ923318.1 | - | 4 | 325 | |||
DQ923319.1 | - | 5 | 425 | |||
DQ923320.1 | - | 4 | 767 |
