siRNA Details
Gene Symbol: Vangl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105969.1 | NP_001099439.1 | 9 | 2007 | ||
XM_006250276.2 | XP_006250338.1 | 10 | 2133 | |||
XM_006250277.2 | XP_006250339.1 | 8 | 1922 |
