siRNA Details
Gene Symbol: Gpr137b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105978.1 | NP_001099448.1 | 5 | 600 | ||
XM_006254372.2 | XP_006254434.1 | 6 | 1102 | |||
XM_006254373.2 | XP_006254435.1 | 6 | 1097 |
