siRNA Details
Gene Symbol: Cr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105989.2 | NP_001099459.2 | 9 | 1510 | ||
XM_006250476.2 | XP_006250538.1 | 13 | 2125 | |||
XM_006250477.2 | XP_006250539.1 | 9 | 1579 | |||
XM_008769847.1 | XP_008768069.1 | 16 | 2710 | |||
XM_008769848.1 | XP_008768070.1 | 9 | 1996 |
