siRNA Details
Gene Symbol: Myo1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134843.1 | NP_001128315.1 | 8, 9 | 1188 | ||
XM_006251218.2 | XP_006251280.1 | 8, 9 | 1203 | |||
XM_006251221.2 | XP_006251283.1 | 8, 9 | 1192 | |||
XM_006251223.2 | XP_006251285.1 | 5, 6 | 582 |
